Läs mer
(-)-alpha-Cedrene (contains (+)-beta-cedrene), TRC
Produktkod. 30351826
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351826 25 g
30364524 5 g
30368191 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30351826 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36486325G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 469-61-4
Molekylformel C15 H24
Molekylvikt (g/mol) 204.35
IUPAC-namn (1S,2R,5S,7S)-2,6,6,8-tetramethyltricyclo[5.3.1.01,5]undec-8-ene
LEDER C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC=C3C
Specifikationer
Kemiskt namn eller material (-)-alpha-Cedrene
Kvantitet 25 g
Synonym alpha-Cedrene, 1H-3a,7-Methanoazulene, 2,3,4,7,8,8a-hexahydro-3,6,8,8-tetramethyl-, (3R,3aS,7S,8aS)-, 1H-3a,7-Methanoazulene, 2,3,4,7,8,8a-hexahydro-3,6,8,8-tetramethyl-, [3R-(3α,3aβ,7β,8aα)]-, Cedr-8-ene (8CI), α-Cedrene (6CI,7CI), (3R,3aS,7S,8aS)-2,3,4,7,8,8a-Hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulene, (-)-Cedrene, (-)-α-Cedrene, Levo-α-cedrene, [3R-(3α,3aβ,7β,8αa)]-2,3,4,7,8,8a-Hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulene
InChI formel InChI=1S/C15H24/c1-10-7-8-15-9-12(10)14(3,4)13(15)6-5-11(15)2/h7,11-13H,5-6,8-9H2,1-4H3/t11-,12+,13+,15+/m1/s1
Formel vikt 204.1878