Läs mer
Alpha-Chlordane, TRC
Produktkod. 30354979
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354979 10 mg
30363542 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30354979 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC32703510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5103-71-9
Molekylformel C10 H6 Cl8
Molekylvikt (g/mol) 409.78
Synonym 4,7-Methano-1H-indene, 1,2,4,5,6,7,8,8-octachloro-2,3,3a,4,7,7a-hexahydro-, (1α,2α,3aα,4β,7β,7aα)-, 4,7-Methanoindan, 1α,2α,4β,5,6,7β,8,8-octachloro-3aα,4,7,7aα-tetrahydro- (8CI), rel-(1R,2S,3aS,4S,7R,7aS)-1,2,4,5,6,7,8,8-Octachloro-2,3,3a,4,7,7a-hexahydro-4,7-methano-1H-indene, (±)-cis-Chlordane, Chlordan, cis-, cis-1,2,4,5,6,7,8,8-Octachloro-3a,4,7,7a-tetrahydro-4,7-endo-methanoindan, cis-Chlordan, cis-Chlordane, cis-α-Chlordane, α-Chlordan, α-Chlordane
IUPAC-namn 1,3,4,7,8,9,10,10-octachlorotricyclo[5.2.1.02,6]dec-8-ene
LEDER Cl[C@H]1C[C@H]2[C@@H]([C@H]1Cl)[C@@]3(Cl)C(=C(Cl)[C@]2(Cl)C3(Cl)Cl)Cl
Specifikationer
Kemiskt namn eller material Cis-Chlordane (Alpha Isomer)
Anteckningar om renhetsgrad GC
Kvantitet 10 mg
InChI formel InChI=1S/C10H6Cl8/c11-3-1-2-4(5(3)12)9(16)7(14)6(13)8(2,15)10(9,17)18/h2-5H,1H2/t2-,3-,4-,5-,8-,9+/m0/s1
Formel vikt 405.7978
Procent renhet >95%