Läs mer
alpha-cis-Bergamotene, TRC
Produktkod. 30337108
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30337108 1 mg
1 options
Produktkod. 30337108 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3184501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (±)-cis-alpha-Bergamotene
CAS 18252-46-5
Molekylformel C15 H24
Kvantitet 1 mg
Synonym alpha-cis-Bergamotene, (±)-cis-alpha-Bergamotene, Bicyclo[3.1.1]hept-2-ene, 2,6-dimethyl-6-(4-methyl-3-penten-1-yl)-, (1R,5R,6R)-rel-, 2-Norpinene, 2,6-dimethyl-6-(4-methyl-3-pentenyl)-, cis- (8CI), Bicyclo[3.1.1]hept-2-ene, 2,6-dimethyl-6-(4-methyl-3-pentenyl)-, (1R,5R,6R)-rel
InChI formel InChI=1S/C15H24/c1-11(2)6-5-9-15(4)13-8-7-12(3)14(15)10-13/h6-7,13-14H,5,8-10H2,1-4H3/t13-,14-,15-/m0/s1
LEDER CC(=CCC[C@@]1(C)[C@H]2CC=C(C)[C@@H]1C2)C
IUPAC-namn (1S,5S,6S)-4,6-dimethyl-6-(4-methylpent-3-enyl)bicyclo[3.1.1]hept-3-ene
Molekylvikt (g/mol) 204.351
Formel vikt 204.188