Läs mer
alpha-Cyclohexyl-benzeneacetic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | alpha-Cyclohexyl-benzeneacetic Acid |
| CAS | 3894-09-5 |
| Molekylformel | C14 H18 O2 |
| Kvantitet | 1 g |
| Synonym | α-Phenyl-cyclohexaneacetic Acid, (±)-Cyclohexylphenylacetic acid, 2-Cyclohexyl-2-phenylacetic acid, Cyclohexylphenyl-acetic Acid, Cyclohexylphenylacetic acid, NSC 60418, α-Cyclohexyl-α-phenylacetic acid, α-Cyclohexylbenzeneacetic acid, α-Cyclohexylphenylacetic acid, α-Phenylcyclohexaneacetic acid |
| InChI formel | InChI=1S/C14H18O2/c15-14(16)13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12-13H,2,5-6,9-10H2,(H,15,16) |
| LEDER | OC(=O)C(C1CCCCC1)c2ccccc2 |
| IUPAC-namn | 2-cyclohexyl-2-phenylacetic acid |
| Molekylvikt (g/mol) | 218.29 |
| Formel vikt | 218.1307 |