Läs mer
alpha-D-Glucopyranuronic Acid Methyl Ester 2,3,4-Tribenzoate 1-Methanesulfonate, TRC
Produktkod. 30408045
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30408045 25 mg
1 options
Produktkod. 30408045 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG45022025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material a-D-Glucopyranuronic Acid Methyl Ester 2,3,4-Tribenzoate 1-Methanesulfonate
CAS 503599-28-8
Molekylformel C29H26O12S
Kvantitet 25 mg
Synonym α-D-Glucopyranuronic acid, methyl ester, 2,3,4-tribenzoate 1-methanesulfonate
InChI formel InChI=1S/C29H26O12S/c1-36-28(33)23-21(37-25(30)18-12-6-3-7-13-18)22(38-26(31)19-14-8-4-9-15-19)24(29(40-23)41-42(2,34)35)39-27(32)20-16-10-5-11-17-20/h3-17,21-24,29H,1-2H3/t21-,22-,23-,24+,29+/m0/s1
LEDER O(C(=O)C1=CC=CC=C1)[C@H]2[C@H](OC(=O)C3=CC=CC=C3)[C@@H](C(OC)=O)O[C@H](OS(C)(=O)=O)[C@@H]2OC(=O)C4=CC=CC=C4
IUPAC-namn (2S,3S,4S,5R,6R)-2-(methoxycarbonyl)-6-((methylsulfonyl)oxy)tetrahydro-2H-pyran-3,4,5-triyl tribenzoate
Molekylvikt (g/mol) 598.57
Formel vikt 598.11