missing translation for 'onlineSavingsMsg'
Läs mer

Alfa Aesar™ alpha-D-Glucose-1-phosphate dipotassium salt hydrate, 98%, Thermo Scientific™

Produktkod. p-7043197
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
10 g
Förpackningsstorlek:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15480507 5 g 5g
15490507 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15480507 Leverantör Alfa Aesar™ Leverantörsnummer J66159.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A α-anomeric form of glucose which contains a phosphate group on the primary carbon. Used as a substance for the estimation of phosphorylases.

Solubility
Soluble in water

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1233690-06-6
Molekylformel C6H17K2O11P
Molekylvikt (g/mol) 374.361
MDL-nummer MFCD00149435
InChI-nyckel RZULLROGARPECB-QMKHLHGBSA-N
Synonym alpha-d-glucopyranose, 1-dihydrogen phosphate , dipotassium salt, dihydrate, alpha-d-glucopyranose 1-phosphate dipotassium salt hydrate, alpha-d-glucose 1-phosphate dipotassium salt dihydrate, alpha-glucose-1-phosphate dipotassium dihydrate, alpha-glucose-1-phosphate dipotassium salt dihydrate mi, a-d-glucose-1-phosphate dipotassium salt hydrate, alpha-d-glucose-1-phosphate dipotassium salt hydrate, dipotassium dihydrate ?-d-glucopyranosyl phosphate, dipotassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphate dihydrate, potassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl phosphate dihydrate
PubChem CID 131855785
IUPAC-namn potassium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate;dihydrate
LEDER C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.O.O.[K].[K]

Specifikationer

Kemiskt namn eller material alpha-D-Glucose-1-phosphate dipotassium salt hydrate
Färg White
Odör Odorless
Kvantitet 5 g
Merck Index 14,4461
Löslighetsinformation Soluble in water
Formel vikt 336.32 (Anhydrous)
Procent renhet 98%
Fysisk form Powder
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.