Läs mer
alpha-D-Maltose Octaacetate, TRC
Produktkod. 30475564
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30475564 500 mg
1 options
Produktkod. 30475564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM158500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-D-Maltose Octaacetate
CAS 6920-00-9
Molekylformel C28H38O19
Kvantitet 500 mg
Synonym 1,2,3,6-Tetra-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl)-alpha-D-glucopyranose
InChI formel InChI=1S/C28H38O19/c1-11(29)37-9-19-21(39-13(3)31)23(40-14(4)32)26(43-17(7)35)28(46-19)47-22-20(10-38-12(2)30)45-27(44-18(8)36)25(42-16(6)34)24(22)41-15(5)33/h19-28H,9-10H2,1-8H3/t19-,20-,21-,22-,23+,24+,25-,26-,27+,28-/m1/s1
LEDER CC(=O)OC[C@H]1O[C@H](OC(=O)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1O[C@H]2O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]2OC(=O)C
IUPAC-namn [(2R,3R,4S,5R,6R)-4,5,6-triacetyloxy-3-[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
Molekylvikt (g/mol) 678.59
Formel vikt 678.2007