missing translation for 'onlineSavingsMsg'
Läs mer

alpha-Demethylnaproxen, Mikromol™

Produktkod. 18081310
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18081310 25 mg
18033410 100 mg
30460728 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18081310 Leverantör Mikromol Leverantörsnummer MM0152.010025

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

alpha-Demethylnaproxen
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material alpha-Demethylnaproxen
CAS 23981-47-7
Analyt- eller komponentnamn alpha-Demethylnaproxen
Frakt skick Room Temperature
Molekylformel C13 H12 O3
Certifikat och efterlevnad ISO 17025
Synonym (6-Methoxynaphthalen-2-yl)acetic Acid, alpha-Demethylnaproxen, Naproxen Imp. I (EP), Naproxen Impurity I
InChI formel InChI=1S/C13H12O3/c1-16-12-5-4-10-6-9(7-13(14)15)2-3-11(10)8-12/h2-6,8H,7H2,1H3,(H,14,15)
LEDER COc1ccc2cc(CC(=O)O)ccc2c1
IUPAC-namn 2-(6-methoxynaphthalen-2-yl)acetic acid
Molekylvikt (g/mol) 216.23
Kvantitet 25 mg
Formel vikt 216.0786 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.