Läs mer
alpha-Demethylnaproxen, Mikromol™
Produktkod. 30460728
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460728 4 x 25 mg
18081310 25 mg
18033410 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460728 Leverantör Mikromol Leverantörsnummer MM0152.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
alpha-Demethylnaproxen, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Demethylnaproxen
CAS 23981-47-7
Analyt- eller komponentnamn alpha-Demethylnaproxen
Frakt skick Room Temperature
Molekylformel C13 H12 O3
Certifikat och efterlevnad ISO 17025
Synonym (6-Methoxynaphthalen-2-yl)acetic Acid, alpha-Demethylnaproxen, Naproxen Imp. I (EP), Naproxen Impurity I
InChI formel InChI=1S/C13H12O3/c1-16-12-5-4-10-6-9(7-13(14)15)2-3-11(10)8-12/h2-6,8H,7H2,1H3,(H,14,15)
Lösningstyp Neat
LEDER COc1ccc2cc(CC(=O)O)ccc2c1
IUPAC-namn 2-(6-methoxynaphthalen-2-yl)acetic acid
Molekylvikt (g/mol) 216.23
Kvantitet 4 x 25 mg
Formel vikt 216.0786 g/mol