Läs mer
(+)-Alpha-Dihydrotetrabenazine, TRC
Produktkod. 30385308
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30385308 25 mg
1 options
Produktkod. 30385308 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45361525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-alpha-Dihydrotetrabenazine
CAS 85081-18-1
Molekylformel C19H29NO3
Kvantitet 25 mg
Synonym (2R,3R,11bR)-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-ol, [2R-(2alpha,3beta,11bbeta)]-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-ol, (+)-(2R,3R,11bR)-Dihydrotetrabenazine, (+)-DTBZ, (+)-Dihydrotetrabenazine, (2R,3R,11bR)-DHTBZ
InChI formel InChI=1S/C19H29NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-9,12,14,16-17,21H,5-7,10-11H2,1-4H3/t14-,16-,17-/m1/s1
LEDER COc1cc2CCN3C[C@@H](CC(C)C)[C@H](O)C[C@@H]3c2cc1OC
IUPAC-namn (2R,3R,11bR)-9,10-dimethoxy-3-(2-methylpropyl)-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizin-2-ol
Molekylvikt (g/mol) 319.44
Formel vikt 319.2147