Läs mer
Alpha-Eleostearic Acid, TRC
Produktkod. 30371856
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30371856 1 mg
1 options
Produktkod. 30371856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8909801MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Alpha-Eleostearic Acid
CAS 506-23-0
Molekylformel C18H30O2
Kvantitet 1 mg
Synonym (9Z,11E,13E)-9,11,13-Octadecatrienoic Acid, cis,trans,trans-9,11,13-Octadecatrienoic Acid, cis-Eleostearic Acid
InChI formel InChI=1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h5-10H,2-4,11-17H2,1H3,(H,19,20)/b6-5+,8-7+,10-9-
LEDER CCCC\C=C\C=C\C=C/CCCCCCCC(=O)O
Molekylvikt (g/mol) 278.43
Formel vikt 278.2246