Learn More
Alpha-Furil, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-Furil |
| CAS | 492-94-4 |
| Molecular Formula | C10H6O4 |
| Quantity | 2.5 g |
| Synonym | 1,2-Di-2-furanyl-1,2-ethanedione, Furil, 1,2-Bis(2-furanyl)-1,2-ethanedione, 1,2-Bis(furan-2-yl)ethanedione, 1,2-Di(2-furanyl)-1,2-ethanedione, 1,2-Di(2-furyl)ethanedione, 1,2-Di(furan-2-yl)ethane-1,2-dione, 1,2-Di-2-furylethane-1,2-dione, 2,2'-Furil, Bipyromucyl, Di-2-furanylethanedione, Difuranylglyoxal, NSC 5561 |
| InChI Formula | InChI=1S/C10H6O4/c11-9(7-3-1-5-13-7)10(12)8-4-2-6-14-8/h1-6H |
| SMILES | O=C(C(=O)c1occc1)c2occc2 |
| Molecular Weight (g/mol) | 190.15 |
| Formula Weight | 190.0266 |