missing translation for 'onlineSavingsMsg'
Läs mer

alpha GalCer:CD1d Complex Monoclonal Antibody (L363), Functional Grade, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-2019-025

 Se fler versioner av denna produkt

Produktkod. 17111192

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

This L363 monoclonal antibody reacts with alpha-galactosylceramide bound to the mouse MHC class I-like protein CD1d. This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Valpha14Jalpha18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation. The L363 antibody has been reported to neutralize iNKT cell activation by alpha-galactosylceramide:CD1d complexes. Applications Reported: This L363 antibody has been reported for use in functional assays. Applications Tested: This L363 antibody has been tested by flow cytometric analysis of alpha-galactosylcermide-loaded mouse CD1d-transfected cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

alpha GalCer:CD1d Complex Monoclonal antibody specifically detects alpha GalCer:CD1d Complex in Mouse samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

alpha GalCer:CD1d Complex
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
P11609, Q9JHE4
Mouse
25 mg
Primary
Mouse
Antibody
IgG2a κ
Flow Cytometry, Functional Assay
L363
Functional Grade
Gal3st1
AI747460; Antigen-presenting glycoprotein CD1d1; Cd1.1; Cd1a; Cd1d; CD1d.1; Cd1d1; CD1d1 antigen; Ly-38; T-cell surface glycoprotein CD1d1
Affinity chromatography
RUO
12479, 53897
4° C
Liquid
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.