Läs mer
alpha-Hydroxy Metoprolol (Mixture of Diastereomers), TRC
Produktkod. 30008480
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008480 5 mg
30007569 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008480 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9483905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56392-16-6
Molekylformel C15 H25 N O4
Molekylvikt (g/mol) 283.363
Synonym Benzenemethanol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-α-(methoxymethyl)- (9CI, ACI), 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-α-(methoxymethyl)benzenemethanol (ACI), 1-Isopropylamino-3-[4-(1-hydroxy-2-methoxyethyl)phenoxy]-2-propanol, H 119/66, α-Hydroxymetoprolol, (R,S)-α-Hydroxymetoprolol, α-Hydroxy Metoprolol, 1-[4-(1-Hydroxy-2-methoxy-ethyl)phenoxy]-3-(isopropylamino)propan-2-ol
IUPAC-namn 1-[4-(1-hydroxy-2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
LEDER COCC(O)c1ccc(OCC(O)CNC(C)C)cc1
Specifikationer
Kemiskt namn eller material a-Hydroxy Metoprolol(Mixture of Diastereomers)
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C15H25NO4/c1-11(2)16-8-13(17)9-20-14-6-4-12(5-7-14)15(18)10-19-3/h4-7,11,13,15-18H,8-10H2,1-3H3
Formel vikt 283.178
Procent renhet >95%