Läs mer
Alpha-(Hydroxyimino)-1H-indole-3-acetic Acid Methyl Ester, TRC
Produktkod. 30407109
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30407109 50 mg
1 options
Produktkod. 30407109 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94328050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-(Hydroxyimino)-1H-indole-3-acetic Acid Methyl Ester
CAS 113975-75-0
Molekylformel C11H10N2O3
Kvantitet 50 mg
Synonym Indole-3-acetic Acid Methyl Ester alpha-Oxime
InChI formel InChI=1S/C11H10N2O3/c1-16-11(14)10(13-15)8-6-12-9-5-3-2-4-7(8)9/h2-6,12,15H,1H3/b13-10+
LEDER COC(=O)\C(=N\O)\c1c[nH]c2ccccc12
IUPAC-namn methyl (2E)-2-hydroxyimino-2-(1H-indol-3-yl)acetate
Molekylvikt (g/mol) 218.21
Formel vikt 218.0691