Läs mer
alpha-Ketoisovaleric Acid, Sodium Salt (13C5), TRC
Produktkod. 30011452
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011452 50 mg
30004869 100 mg
30011809 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011452 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK54833050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173018-24-0
Molekylformel C5H7NaO3
Molekylvikt (g/mol) 143.06
Synonym α-Keto Isovaleric Acid Sodium Salt 13C5
LEDER [Na+].[13CH3][13CH]([13CH3])[13C](=O)[13C](=O)[O-]
Specifikationer
Kemiskt namn eller material alpha-Ketoisovaleric Acid, Sodium Salt (13C5)
Kvantitet 50 mg
InChI formel InChI=1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1/i1+1,2+1,3+1,4+1,5+1;