Läs mer
(±)-alpha-Lipoamide, TRC
Produktkod. 30138527
Change view
Kvantitet:
1 g
2.5 g
10 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138527 2.5 g 2.5g
30133024 10 g -
30009550 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138527 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4687102.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 940-69-2
Molekylformel C8H15NOS2
Molekylvikt (g/mol) 205.34
Synonym 1,2-Dithiolane-3-pentanamide, 1,2-Dithiolane-3-valeramide, α-Lipamide, (±)-1,2-Dithiolane-3-pentanamide, (±)-1,2-Dithiolane-3-valeramide, (±)-Thioctic Acid Amide, 5-(1,2-Dithiolan-3-yl)pentanamide, 5-(1,2-Dithiolan-3-yl)valeramide, DL-6,8-Thioctamide, DL-Lipoamide, DL-α-Lipoic Acid Amide, Lipamide, Lipoacin, Lipoamid, Lipoamide, Lipoicin, NSC 90787, Pathoclon, Thioami, Thioctamid, Thioctamide, Thioctic Acid Amide, Thioctic Amide, Thiotomin, Ticolin, Vitamin N, α-Lipoamide;
LEDER NC(CCCCC1CCSS1)=O
Specifikationer
Kemiskt namn eller material (±)-alpha-Lipoamide
Kvantitet 2.5 g
InChI formel InChI=1S/C8H15NOS2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H2,9,10)