Läs mer
alpha-Lobeline Hydrochcloride, TRC
Produktkod. 30472268
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472268 500 mg
30475460 100 mg
30471346 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472268 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL469350500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 134-63-4
Molekylformel C22 H27 N O2 . Cl H
Molekylvikt (g/mol) 373.92
Synonym Lobeline hydrochloride, 2-[(2R,6S)-6-[(2S)-2-Hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone hydrochloride
IUPAC-namn 2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;hydrochloride
LEDER Cl.CN1[C@H](C[C@H](O)c2ccccc2)CCC[C@@H]1CC(=O)c3ccccc3
Specifikationer
Kemiskt namn eller material alpha-Lobeline hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C22H27NO2.ClH/c1-23-19(15-21(24)17-9-4-2-5-10-17)13-8-14-20(23)16-22(25)18-11-6-3-7-12-18;/h2-7,9-12,19-21,24H,8,13-16H2,1H3;1H/t19-,20+,21-;/m0./s1
Formel vikt 373.1809