Läs mer
Alpha-Methyl-D,L-p-tyrosine Methyl Ester Hydrochloride, TRC
Produktkod. 30475596
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30475596 5 mg
1 options
Produktkod. 30475596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3327405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material a-Methyl-D,L-p-tyrosine, Methyl Ester, Hydrochloride
CAS 7361-31-1
Molekylformel C11 H15 N O3 . Cl H
Kvantitet 5 mg
InChI formel InChI=1S/C11H15NO3.ClH/c1-11(12,10(14)15-2)7-8-3-5-9(13)6-4-8;/h3-6,13H,7,12H2,1-2H3;1H
LEDER Cl.COC(=O)C(C)(N)Cc1ccc(O)cc1
IUPAC-namn methyl 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate;hydrochloride
Molekylvikt (g/mol) 245.7
Formel vikt 245.0819