Läs mer
alpha-Methyl-D-serine, TRC
Produktkod. 30479130
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479130 50 mg
30482853 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479130 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32597550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81132-44-7
Molekylformel C4 H9 N O3
Molekylvikt (g/mol) 119.12
IUPAC-namn (2R)-2-amino-3-hydroxy-2-methylpropanoic acid
LEDER C[C@@](N)(CO)C(=O)O
Specifikationer
Kemiskt namn eller material a-Methyl-D-serine
InChI formel InChI=1S/C4H9NO3/c1-4(5,2-6)3(7)8/h6H,2,5H2,1H3,(H,7,8)/t4-/m1/s1
Kvantitet 50 mg
Formel vikt 119.0582