Läs mer
Alpha-Methyl-D-tryptophan, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | a-Methyl-D-tryptophan |
| CAS | 56452-52-9 |
| Molekylformel | C12 H14 N2 O2 |
| Kvantitet | 25 mg |
| Synonym | D-Tryptophan, α-methyl-, α-Methyl-D-tryptophan, (+)-α-Methyltryptophan, (R)-2-Amino-3-(1H-indol-3-yl)-2-methylpropanoic acid, (R)-α-Methyltryptophan, D-α-Methyltryptophan, α-Methyl-(R)-tryptophan |
| InChI formel | InChI=1S/C12H14N2O2/c1-12(13,11(15)16)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,14H,6,13H2,1H3,(H,15,16)/t12-/m1/s1 |
| LEDER | C[C@@](N)(Cc1c[nH]c2ccccc12)C(=O)O |
| IUPAC-namn | (2R)-2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid |
| Molekylvikt (g/mol) | 218.25 |
| Formel vikt | 218.1055 |