Läs mer
alpha-Methyl-DL-tryptophan, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | alpha-Methyl-DL-tryptophan |
| CAS | 153-91-3 |
| Molekylformel | C12 H14 N2 O2 |
| Synonym | Tryptophan, α-methyl-, DL-Tryptophan, α-methyl-, Tryptophan, α-methyl-, DL- (8CI), α-Methyltryptophan, (±)-α-Methyltryptophan, 2-Amino-3-(1H-indol-3-yl)-2-methylpropanoic acid, 2-Azaniumyl-3-(1H-indol-3-yl)-2-methylpropanoate, DL-α-Methyltryptophan, NSC 9948, α-Methyl-DL-tryptophan |
| InChI formel | InChI=1S/C12H14N2O2/c1-12(13,11(15)16)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,14H,6,13H2,1H3,(H,15,16) |
| LEDER | CC(N)(Cc1c[nH]c2ccccc12)C(=O)O |
| IUPAC-namn | 2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid |
| Molekylvikt (g/mol) | 218.25 |
| Kvantitet | 1 g |
| Formel vikt | 218.1055 |
| Visa mer |