Läs mer
Alpha-Methyl-L-valine, TRC
Produktkod. 30474073
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30474073 500 mg
1 options
Produktkod. 30474073 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338540500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material a-Methyl-L-valine
CAS 53940-83-3
Molekylformel C6 H13 N O2
Kvantitet 500 mg
Synonym L-Valine, 2-methyl-, L-Isovaline, 3-methyl- (9CI), 2-Methyl-L-valine, (S)-2-Methylvaline, (S)-α-Methylvaline, L-α-Methylvaline
InChI formel InChI=1S/C6H13NO2/c1-4(2)6(3,7)5(8)9/h4H,7H2,1-3H3,(H,8,9)/t6-/m0/s1
LEDER CC(C)[C@](C)(N)C(=O)O
IUPAC-namn (2S)-2-amino-2,3-dimethylbutanoic acid
Molekylvikt (g/mol) 131.17
Formel vikt 131.0946