Läs mer
Alpha-Methyl-trans-cinnamaldehyde, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | alpha-Methyl-trans-cinnamaldehyde |
| CAS | 15174-47-7 |
| Molekylformel | C10H10O |
| Kvantitet | 25 g |
| Synonym | 2-Methyl-3-phenyl-2-propenal, 2-Methyl-3-phenyl-2-propen-1-al, 2-Methyl-3-phenylacrolein, 2-Methyl-3-phenylacrylaldehyde, 2-Methyl-3-phenylpropenal, 2-Methylcinnamic Aldehyde, Methylcinnamaldehyde, NSC 22283, NSC 49286, alpha-Methylcinnamaldehyde, alpha-Methylcinnamic Aldehyde, alpha-Methyl-cinnamaldehyde |
| InChI formel | InChI=1S/C10H10O/c1-9(8-11)7-10-5-3-2-4-6-10/h2-8H,1H3/b9-7+ |
| LEDER | C\C(=C/c1ccccc1)\C=O |
| IUPAC-namn | (E)-2-methyl-3-phenylprop-2-enal |
| Molekylvikt (g/mol) | 146.19 |
| Formel vikt | 146.0732 |