Läs mer
Alpha-Methyldopa Methyl Ester, TRC
Produktkod. 30180251
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180251 5 mg
30179731 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180251 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3038635MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18181-08-3
Molekylformel C11 H15 N O4
Molekylvikt (g/mol) 225.24
Synonym Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methyl-propanoate, 3-Hydroxy-α-methyl-L-tyrosine Methyl Ester, Alanine, 3-(3,4-dihydroxyphenyl)-2-methyl-, methyl ester, L- (8CI), α-Methyldopa Methyl Ester
IUPAC-namn methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate
LEDER COC(=O)[C@@](C)(N)Cc1ccc(O)c(O)c1
Specifikationer
Kemiskt namn eller material Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate
Kvantitet 5 mg
InChI formel InChI=1S/C11H15NO4/c1-11(12,10(15)16-2)6-7-3-4-8(13)9(14)5-7/h3-5,13-14H,6,12H2,1-2H3/t11-/m0/s1
Formel vikt 225.1001