Läs mer
alpha-Methyltryptamine Succinate (AMT Succinate, alphaMT Succinate), LoGiCal
Specifikationer
Specifikationer
| Kemiskt namn eller material | alpha-Methyltryptamine Succinate (AMT Succinate, alphaMT Succinate) |
| CAS | 856648-01-6 |
| Analyt- eller komponentnamn | alpha-Methyltryptamine Succinate (AMT Succinate, alphaMT Succinate) |
| Frakt skick | Room Temperature |
| Molekylformel | C11 H14 N2 . C4 H6 O4 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | alpha-Methyltryptamine Succinate, AMT Succinate, alphaMT Succinate, α-Methyltryptamine Succinate, αMT Succinate, Butanedioic acid compd. with α-methyl-1H-indole-3-ethanamine (1:1), Succinic acid compd. with 3-(2-aminopropyl)indole |
| InChI formel | InChI=1S/C11H14N2.C4H6O4/c1-8(12)6-9-7-13-11-5-3-2-4-10(9)11;5-3(6)1-2-4(7)8/h2-5,7-8,13H,6,12H2,1H3;1-2H2,(H,5,6)(H,7,8) |
| Lösningstyp | Neat |
| LEDER | CC(N)Cc1c[nH]c2ccccc12.OC(=O)CCC(=O)O |
| Visa mer |