Läs mer
alpha-Phenylethylamine Hydrochloride, Mikromol
Beskrivning
alpha-Phenylethylamine Hydrochloride, Mikromol
Specifikationer
Specifikationer
| Kemiskt namn eller material | alpha-Phenylethylamine Hydrochloride |
| CAS | 13437-79-1 |
| Analyt- eller komponentnamn | alpha-Phenylethylamine Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C8 H11 N . H Cl |
| Synonym | α-Phenylethylamine Hydrochloride, Benzenemethanamine, α-methyl-, hydrochloride (1:1), Benzenemethanamine, α-methyl-, hydrochloride (9CI), Benzenemethanamine, α-methyl-, hydrochloride, (±)-, Benzylamine, α-methyl-, hydrochloride, (±)- (8CI), (±)-1-Phenylethylamine hydrochloride, (±)-α-Methylbenzylamine hydrochloride, 1-Phenylethylamine hydrochloride, dl-1-Phenylethylammonium chloride, α-Methylbenzylamine hydrochloride |
| InChI formel | InChI=1S/C8H11N.ClH/c1-7(9)8-5-3-2-4-6-8;/h2-7H,9H2,1H3;1H |
| Lösningstyp | Neat |
| LEDER | Cl.CC(N)c1ccccc1 |
| IUPAC-namn | 1-phenylethanamine;hydrochloride |
| Visa mer |