Läs mer
Alpha-Phenyllongifolol, TRC
Produktkod. 30426119
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30426119 10 mg
1 options
Produktkod. 30426119 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP33553010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Phenyllongifolol
CAS 946513-19-5
Molekylformel C21H30O
Kvantitet 10 mg
Synonym (αR,1S,3aR,4S,8aS,9R)-Decahydro-4,8,8-trimethyl-α-phenyl-1,4-methanoazulene-9-methanol
InChI formel InChI=1S/C26H27ClN2O3S2.Na/c1-29(2)24(30)12-14-33-26(34-15-13-25(31)32)20-5-3-4-18(16-20)6-10-22-11-8-19-7-9-21(27)17-23(19)28-22;/h3-11,16-17,26H,12-15H2,1-2H3,(H,31,32);/q;+1/p-1/b10-6+;
LEDER [Na+].O=C([O-])CCSC(SCCC(=O)N(C)C)c1cc(ccc1)/C=C/c2nc3c(ccc(Cl)c3)cc2
Molekylvikt (g/mol) 298.46