Läs mer
Alpha-Pinene Oxide, TRC
Produktkod. 30412200
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30412200 500 mg
1 options
Produktkod. 30412200 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP466620500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Pinene Oxide
CAS 1686-14-2
Molekylformel C10 H16 O
Kvantitet 500 mg
Synonym 2,3-Epoxypinane, 2,7,7-trimethyl-3-oxatricyclo[4.1.1.02,4]octane
InChI formel InChI=1S/C10H16O/c1-9(2)6-4-7(9)10(3)8(5-6)11-10/h6-8H,4-5H2,1-3H3
LEDER CC1(C)C2CC3OC3(C)C1C2
Molekylvikt (g/mol) 152.23
Formel vikt 152.1201