Läs mer
alpha-Tocopherol Phosphate Disodium Salt, TRC
Produktkod. 30452460
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30452460 50 mg
1 options
Produktkod. 30452460 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT52618550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Tocopherol Phosphate Disodium Salt
CAS 90940-45-7
Molekylformel C29 H49 Na2 O5 P
Kvantitet 50 mg
Synonym α-Tocopherol Phosphate Disodium Salt
InChI formel InChI=1S/C29H51O5P.2Na/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(34-35(30,31)32)23(5)24(6)28(26)33-29;;/h20-22H,9-19H2,1-8H3,(H2,30,31,32);;/q;2*+1/p-2/t21-,22-,29-;;/m1../s1
LEDER CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OP(=O)(O[Na])O[Na])c(C)c(C)c2O1
IUPAC-namn [sodiooxy-[[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl]oxy]phosphoryl]oxysodium
Molekylvikt (g/mol) 554.65
Formel vikt 554.311