Läs mer
Alpha-Zearalenol, TRC
Produktkod. 30041359
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30041359 0.5 mg 0.5mg
30041333 5 mg -
30041097 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041359 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ2704700.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36455-72-8
Molekylformel C18 H24 O5
Molekylvikt (g/mol) 320.38
Synonym 1H-2-Benzoxacyclotetradecin-1-one, 3,4,5,6,7,8,9,10-octahydro-7,14,16-trihydroxy-3-methyl-, [3S-(3R*,7S*,11E)]-, (3S,7R,11E)-3,4,5,6,7,8,9,10-Octahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one, (-)-α-Zearalenol, Zearalenol, trans-Zearalenol, α-Zearalenol
IUPAC-namn (2E,7R,11S)-7,15,17-trihydroxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(18),2,14,16-tetraen-13-one
LEDER C[C@H]1CCC[C@H](O)CCC\C=C\c2cc(O)cc(O)c2C(=O)O1
Specifikationer
Kemiskt namn eller material alpha-Zearalenol
Anteckningar om renhetsgrad HPLC
Kvantitet 0.5 mg
InChI formel InChI=1S/C18H24O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,14,19-21H,2,4-6,8-9H2,1H3/b7-3+/t12-,14+/m0/s1
Formel vikt 320.1624
Procent renhet >95%