missing translation for 'onlineSavingsMsg'
Learn More

Alphacel Non-Nutritive Bulk, MP Biomedicals

Catalog Number p-3545911
Click to view available options
Quantity:
5 lb.
25 lb.
100 lb.
This item is not returnable. View return policy
9004-34-6
This item is not returnable. View return policy
9004-34-6

Composed of finely ground cellulose for addition to animal diets to supply bulk with minimum nutritional value

Quantity 5 lb.
Chemical Name or Material Alphacel Non-Nutritive Bulk
compliance-icons
Product Identifier
  • alphacel
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : alphacel
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'