Läs mer
(AlphaE)-2-[(6-Chloro-4-pyrimidinyl)oxy]-Alpha-(methoxymethylene)benzeneacetic Acid Methyl Ester, TRC
Produktkod. 30350204
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30350204 10 mg
1 options
Produktkod. 30350204 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38024010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (alphaE)-2-[(6-Chloro-4-pyrimidinyl)oxy]-alpha-(methoxymethylene)benzeneacetic Acid Methyl Ester
CAS 131860-97-4
Molekylformel C15H13ClN2O4
Kvantitet 10 mg
Synonym (E)-2-[(6-Chloro-4-pyrimidinyl)oxy]-alpha-(methoxymethylene)benzeneacetic Acid Methyl Este, Methyl (E)-2-[2-(6-Chloropyrimidin-4-yloxy)phenyl]-3-methoxyacrylate, Methyl (E)-2-[2-(6-Chloropyrimidin-4-yloxy)phenyl]-3-methoxypropenoate
InChI formel InChI=1S/C15H13ClN2O4/c1-20-8-11(15(19)21-2)10-5-3-4-6-12(10)22-14-7-13(16)17-9-18-14/h3-9H,1-2H3/b11-8+
LEDER CO\C=C(\C(=O)OC)/c1ccccc1Oc2cc(Cl)ncn2
IUPAC-namn methyl (E)-2-[2-(6-chloropyrimidin-4-yl)oxyphenyl]-3-methoxyprop-2-enoate
Molekylvikt (g/mol) 320.73
Formel vikt 320.0564