Läs mer
(AlphaR)-Alpha-Methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-2-naphthalenemethanamine Hydrochloride, TRC
Produktkod. 30475046
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30475046 100 mg
1 options
Produktkod. 30475046 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM331105100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (alphaR)-alpha-Methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-2-naphthalenemethanamine Hydrochloride
CAS 2200269-28-7
Molekylformel C22H22F3N . HCl
Kvantitet 100 mg
Synonym 2-Naphthalenemethanamine, α-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride (1:1), (αR)- (ACI), (αR)-α-Methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-2-naphthalenemethanamine hydrochloride, N-[(1R)-1-(2-Naphthyl)ethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine hydrochloride, [(1R)-1-(Naphthalen-2-yl)ethyl]([3-[3-(trifluoromethyl)phenyl]propyl])amine hydrochloride
InChI formel InChI=1S/C22H22F3N.ClH/c1-16(19-12-11-18-8-2-3-9-20(18)15-19)26-13-5-7-17-6-4-10-21(14-17)22(23,24)25;/h2-4,6,8-12,14-16,26H,5,7,13H2,1H3;1H/t16-;/m1./s1
LEDER Cl.C[C@@H](NCCCc1cccc(c1)C(F)(F)F)c2ccc3ccccc3c2
IUPAC-namn N-[(1R)-1-naphthalen-2-ylethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine;hydrochloride
Molekylvikt (g/mol) 393.9
Formel vikt 393.1471