Läs mer
(AlphaR,BetaS)-rel-Bedaquiline, TRC
Produktkod. 30339982
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30339982 10 mg
1 options
Produktkod. 30339982 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB11955510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (alphaR,betaS)-rel-Bedaquiline
CAS 654653-93-7
Molekylformel C32H31BrN2O2
Kvantitet 10 mg
Synonym (aR,bS)-rel-Bedaquiline
InChI formel InChI=1S/C32H31BrN2O2/c1-35(2)19-18-32(36,28-15-9-13-22-10-7-8-14-26(22)28)30(23-11-5-4-6-12-23)27-21-24-20-25(33)16-17-29(24)34-31(27)37-3/h4-17,20-21,30,36H,18-19H2,1-3H3/t30?,32-/m1/s1
LEDER BrC1=CC=C2C(C=C([C@@H](C3=CC=CC=C3)[C@@](O)(CCN(C)C)C4=C(C=CC=C5)C5=CC=C4)C(OC)=N2)=C1
IUPAC-namn (1R,2S)-1-(6-bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol
Molekylvikt (g/mol) 555.5
Formel vikt 554.16