Läs mer
(alphaS)-alpha-Amino-N-[(1R)-1-[(3aS,4S,6S,7aR)-hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl]-3-methylbutyl]benzenepropanamide Hydrochloride (Bortezomib Intermediate II), Mikromol™
Produktkod. 30050881
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30050881 25 mg
1 options
Produktkod. 30050881 Leverantör Mikromol Leverantörsnummer MM3368.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(alphaS)-alpha-Amino-N-[(1R)-1-[(3aS,4S,6S,7aR)-hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl]-3-methylbutyl]benzenepropanamide Hydrochloride (Bortezomib Intermediate II)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (alphaS)-alpha-Amino-N-[(1R)-1-[(3aS,4S,6S,7aR)-hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl]-3-methylbutyl]benzenepropanamide Hydrochloride
CAS 205393-21-1
Analyt- eller komponentnamn (alphaS)-alpha-Amino-N-[(1R)-1-[(3aS,4S,6S,7aR)-hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl]-3-methylbutyl]benzenepropanamide Hydrochloride
Frakt skick Room Temperature
Molekylformel C24 H37 B N2 O3 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Benzenepropanamide, α-amino-N-[(1R)-1-[(3aS,4S,6S,7aR)-hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl]-3-methylbutyl]-, hydrochloride (1:1), (αS)-, Benzenepropanamide, α-amino-N-[(1R)-1-[(3aS,4S,6S,7aR)-hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl]-3-methylbutyl]-, monohydrochloride, (αS)- (9CI), Benzenepropanamide, α-amino-N-[1-(hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl)-3-methylbutyl]-, monohydrochloride, [3aS-[2[S*(R*)],3aα,4β,6β,7aα]]-
InChI formel InChI=1S/C24H37BN2O3.ClH/c1-15(2)11-21(27-22(28)18(26)12-16-9-7-6-8-10-16)25-29-20-14-17-13-19(23(17,3)4)24(20,5)30-25;/h6-10,15,17-21H,11-14,26H2,1-5H3,(H,27,28);1H/t17-,18-,19-,20+,21-,24-;/m0./s1
LEDER Cl.CC(C)C[C@H](NC(=O)[C@@H](N)Cc1ccccc1)B2O[C@@H]3C[C@@H]4C[C@@H](C4(C)C)[C@]3(C)O2
Molekylvikt (g/mol) 448.8341
Kvantitet 25 mg
Formel vikt 448.2664 g/mol