Läs mer
Skytec™ Alphashield 2000+- S2BH+ Disposable Coverall
Storlek:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Beskrivning
- Spun bonded polypropylene with outer breathable film with chemical liquid (EN368 and blood (ASTM F1670) penetration resistance for premium protection
- Strong, durable, and comfortable
- Improved fit with two-piece crotch pattern
- Quick and easy donning/doffing in an emergency with longer 2-way zip
- Fully elasticated hood, ankles, and wrists
Specifikationer
Specifikationer
| Produkttyp | Chemical-resistant Coveralls |
| Storlek | 3X-Large |
| Kön | Unisex |
| Material | Polypropylene |
| Färg | White |
| Disponibel | Single-use |
| Midja stil | Elastic |
| Ankel stil | Elastic |
| Manschettstil | Elastic |
| Hals stil | Hooded |
| Visa mer |