Learn More
Ansell™ AlphaTec™ 3000 Oversleeves Ultrasonically Welded and Taped
Product Code. 31114790 Shop All Ansell Products
Change view
Click to view available options
Style:
Elastic at Both Ends
Product Code. Style
31114790 Elastic at Both Ends
1 options
Product Code. 31114790 Supplier Ansell™ Supplier No. YE30T0060000
Beräknad leverans: 10-12-2026
Multi-layer barrier fabric for protection against hazardous chemicals with improved personal protection
  • The AlphaTec™ 3000 Oversleeves Ultrasonically Welded and Taped - Model 600 is an excellent addition to any chemical protective suit, as they are made of multi-layer barrier fabric that provides reliable protection against a wide range of chemicals
  • These protective arm sleeves are in bright yellow, granting high visibility for improved workplace safety, particularly in low-light environments, besides providing durable protection against accidents and injuries
  • The ultrasonically welded and taped seams of these forearm protection sleeves offer high levels of durability, ensuring no tearing and ripping
TRUSTED_SUSTAINABILITY
Specifications
Size Standard
Product Type Chemical-resistant Sleeve
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Sterility Non-sterile
Style Elastic at Both Ends
Disposable Single-use
Seam Type Ultrasonically Welded
Latex Free Yes
Certifications/Compliance CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126