Learn More
Ansell AlphaTec™ 39-035 Cotton-Supported Nitrile Chemical Protection Gloves
Robust gloves offer a high level of resistance against animal fats, tears, and abrasions.
Brand: Ansell 39035090
Additional Details : Weight : 14.11200kg
Description
- Industrial protective gloves
- Supported cotton construction
- Smooth finish for dry grip applications
- High level of resistance to animal fats
Specifications
EN 374_2003 Microorganism, EN 420:2003 + A1:2009 | |
No Texture | |
9 | |
Type A | |
Yes | |
39-035 |
Green | |
Nitrile | |
3121A | |
JKLOPT | |
Gauntlet |