Läs mer
(alphaZ)-alpha-(Methoxyimino)-N-[(5aR,6R)-1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl]-2-furanacetamide, TRC
Produktkod. 30137664
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137664 50 mg
17971325 100 mg
17952655 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26274550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 947723-87-7
Synonym (αZ)-α-(Methoxyimino)-N-[(5aR,6R)-1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl]-2-furanacetamide∣Cefuroxime Imp. H (EP)∣Cefuroxime Axetil Imp. E (EP)
Specifikationer
Formatera Neat
Molekylvikt (g/mol) 363.35
Kvantitet 50 mg
Frakt skick Blue Ice