missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ALR |
---|---|
Concentration | 0.36 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828273
|
Proteintech
11293-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818273
|
Proteintech
11293-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Isoform 1: FAD-dependent sulfhydryl oxidase that regenerates the redox-active disulfide bonds in CHCHD4/MIA40, a chaperone essential for disulfide bond formation and protein folding in the mitochondrial intermembrane space. The reduced form of CHCHD4/MIA40 forms a transient intermolecular disulfide bridge with GFER/ERV1, resulting in regeneration of the essential disulfide bonds in CHCHD4/MIA40, while GFER/ERV1 becomes re- oxidized by donating electrons to cytochrome c or molecular oxygen.Specifications
ALR | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P55789, P56213, Q63042 | |
11692, 2671, 27100 | |
ALR Fusion Protein Ag1840 | |
Primary | |
-20°C | |
GFER |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ALR, ERV1, GFER, Hepatopoietin, HERV1, HPO, HPO1, HPO2, HSS | |
Gfer | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |