Läs mer
Alternariol 9-Methyl Ether 3-Sulfate Ammonium Salt, TRC
Produktkod. 30325614
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30325614 10 mg
1 options
Produktkod. 30325614 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA57567010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Alternariol 9-Methyl Ether 3-Sulfate Ammonium Salt
CAS 1488351-25-2
Molekylformel C15H12O8S(NH3)
Kvantitet 10 mg
Synonym Alternariol Monomethyl Ether 3-Sulfate Ammonium Salt, 7-Hydroxy-9-methoxy-1-methyl-3-(sulfooxy)-6H-dibenzo[b,d]pyran-6-one Ammonium Salt
InChI formel InChI=1S/C15H12O8S.H3N/c1-7-3-9(23-24(18,19)20)6-12-13(7)10-4-8(21-2)5-11(16)14(10)15(17)22-12;/h3-6,16H,1-2H3,(H,18,19,20);1H3
LEDER CC1=C2C(OC(C3=C2C=C(OC)C=C3O)=O)=CC(OS(O)(=O)=O)=C1.N
IUPAC-namn 7-hydroxy-9-methoxy-1-methyl-6-oxo-6H-benzo[c]chromen-3-yl hydrogen sulfate ammonium salt
Molekylvikt (g/mol) 369.35
Formel vikt 369.06