Läs mer
Alternariol Monomethyl Ether, TRC
Produktkod. 18002920
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18002920 1 mg
18052310 5 mg
18023920 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18002920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5757701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 26894-49-5
Molekylformel C15 H12 O5
Molekylvikt (g/mol) 272.253
Synonym 2-Biphenylcarboxylic acid, 2',3,4'-trihydroxy-5-methoxy-6'-methyl-, δ-lactone (6CI), 3,7-Dihydroxy-9-methoxy-1-methyl-6H-dibenzo[b,d]pyran-6-one, AME, Alternariol 9-O-methyl ether, Alternariol 9-methyl ether, Alternariol monomethyl ether, Alternariol-5-O-methyl ether, Djalonensone, NSC 638262
IUPAC-namn 3,7-dihydroxy-9-methoxy-1-methylbenzo[c]chromen-6-one
LEDER COc1cc(O)c2C(=O)Oc3cc(O)cc(C)c3c2c1
Specifikationer
Kemiskt namn eller material Alternariolmethylether
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C15H12O5/c1-7-3-8(16)4-12-13(7)10-5-9(19-2)6-11(17)14(10)15(18)20-12/h3-6,16-17H,1-2H3
Formel vikt 272.068
Procent renhet >95%