Läs mer
Altretamine, TRC
Produktkod. 30320868
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30320868 5 g
1 options
Produktkod. 30320868 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5757655G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Altretamine
CAS 645-05-6
Molekylformel C9 H18 N6
Kvantitet 5 g
Synonym 1,3,5-Triazine-2,4,6-triamine, N2,N2,N4,N4,N6,N6-hexamethyl-, 1,3,5-Triazine-2,4,6-triamine, N,N,N',N',N'',N''-hexamethyl- (9CI), Melamine, hexamethyl- (7CI,8CI), N2,N2,N4,N4,N6,N6-Hexamethyl-1,3,5-triazine-2,4,6-triamine, 1,3,5-Tris(dimethylamino)-s-triazine, 2,4,6-Tris(dimethylamino)-1,3,5-triazine, 2,4,6-Tris(dimethylamino)-s-triazine, Altretamine, ENT 50852, HMM, Hemel, Hexalen, Hexamethylmelamine, Hexylen, NSC 13875
InChI formel InChI=1S/C9H18N6/c1-13(2)7-10-8(14(3)4)12-9(11-7)15(5)6/h1-6H3
LEDER CN(C)c1nc(nc(n1)N(C)C)N(C)C
IUPAC-namn 2-N,2-N,4-N,4-N,6-N,6-N-hexamethyl-1,3,5-triazine-2,4,6-triamine
Molekylvikt (g/mol) 210.28
Formel vikt 210.1593