missing translation for 'onlineSavingsMsg'
Learn More

Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50, Thermo Scientific Chemicals

Catalog Number p-4509878
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
12635-29-9
AlNi
85.675
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
6336846
aluminum;nickel
[Al].[Ni]
12635-29-9
AlNi
85.675
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
6336846
aluminum;nickel
[Al].[Ni]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black-Gray to Gray
Melting Point 1350.0°C
Density 3.4600g/mL
Quantity 100 g
Fieser 01,723; 02,293; 05,570; 06,502; 07,312; 01,723; 02,293; 05,570; 06,502; 07,312; 08,433; 09,405; 10,339; 11,457; 12,422; 13,265; 14,270; 15,278; 17,296
Formula Weight 85.68
Packaging Glass bottle
Form Powder
Chemical Name or Material Aluminum-nickel
compliance-icons compliance-icons
Product Identifier
  • Aluminium-nickel, Raney-type alloy, powder
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H351-Suspected of causing cancer.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'