missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, for Dioxin Analysis, Thermo Scientific™

Catalog Number p-7080042
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
dialuminium(3+) trioxidandiide
[O--].[O--].[O--].[Al+3].[Al+3]
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
dialuminium(3+) trioxidandiide
[O--].[O--].[O--].[Al+3].[Al+3]

For Dioxin Analysis

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 2980°C
Melting Point 2045°C
Color White
Physical Form Powder
Quantity 250 g
Linear Formula Al2O3
Merck Index 14,356
Formula Weight 101.96
Odor Odorless
Refractive Index 1.765
Chemical Name or Material Aluminum oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'