Learn More
Aluminum oxide, for TLC, with gypsum, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Boiling Point | 2980°C |
Melting Point | 2045°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Merck Index | 14,356 |
Formula Weight | 101.96 |
Odor | Odorless |
Refractive Index | 1.765 |
Chemical Name or Material | Aluminum oxide, With gypsum |