Learn More
Reichert Instruments™ Aluminum Reichert Refractometer Table Stand
Brand: Reichert Instruments™ 13104080
Additional Details : Weight : 0.80000kg
Specifications
Reichert Refractometer Table Stand | |
EU Cord | |
230 V 50 Hz | |
50 Hz |
TS meter illuminated | |
1 Each | |
230 V |