Läs mer
VACUUBRAND™ T-stycke i aluminium med små KF-flänsar
Produktkod. 10039560 Handla allt Vacuubrand Produkter
Change view
Diameter (metrisk) Insida:
10/10/10 mm
16/16/16 mm
25/25/25 mm
40/40/40 mm
Storlek:
30 x 60 mm
40 x 80 mm
50 x 100 mm
65 x 130 mm
Förpackningsstorlek:
Varje
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Diameter (metrisk) Insida Storlek unitSize
10039560 10/10/10 mm 30 x 60 mm -
10273371 16/16/16 mm 40 x 80 mm Varje
10450273 25/25/25 mm 50 x 100 mm Varje
10589911 40/40/40 mm 65 x 130 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 10039560 Leverantör VACUUBRAND™ Leverantörsnummer 20669500