Läs mer
Alvelestat, MedChemExpress
Produktkod. 16496718
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496718 10 mg
16412807 50 mg
16409277 5 mg
16406866 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496718 Leverantör Medchem Express Leverantörsnummer HY15651
MedChemExpress Alvelestat (AZD9668) is an orally bioavailable, affinity and selective inhibitor of neutrophil elastase (NE) with a pIC50 value of 7.9 nM, a Ki value of 9.4 nM and a Kd value of 9.5 nM.
Alvelestat (AZD9668) is an orally bioavailable, affinity and selective inhibitor of neutrophil elastase (NE) with a pIC50 value of 7.9 nM, a Ki value of 9.4 nM and a Kd value of 9.5 nM.
Specifikationer
Kemiskt namn eller material Alvelestat
Fysisk form Solid
CAS 848141-11-7
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C25H22F3N5O4S
För användning med (applikation) COVID-19-immunoregulation
Synonym AZD9668
Löslighetsinformation DMSO : ≥ 33 mg/mL (60.49 mM)
LEDER O=C(C1=CC(C2=CC=NN2C)=C(C)N(C3=CC=CC(C(F)(F)F)=C3)C1=O)NCC4=NC=C(S(=O)(C)=O)C=C4
Molekylvikt (g/mol) 545.53
Formel vikt 545.53
Procent renhet 95.43%